missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzophenone, 97%

Manufacturer:  Alfa Aesar™ H5350506

 View more versions of this product

Catalog No. AAH5350506



Description

Description

Specifications

Specifications

3-Bromobenzophenone
5g
C13H9BrO
3-bromobenzophenone, 3-bromobenzoylbenzene, 3-bromophenyl phenyl ketone, 3-bromophenyl phenyl methanone, 3-bromophenyl phenylmethanone, 3-bromophenyl-phenyl-methanone, acmc-20an8e, bromobenzophenone,m, m-bromobenzophenone, methanone, 3-bromophenyl phenyl
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Br
261.118
261.12
74°C to 78°C
1016-77-9
MFCD00672015
XNUMUNIJQMSNNN-UHFFFAOYSA-N
(3-bromophenyl)-phenylmethanone
70548
97%

Chemical Identifiers

1016-77-9
261.118
XNUMUNIJQMSNNN-UHFFFAOYSA-N
70548
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Br
C13H9BrO
MFCD00672015
3-bromobenzophenone, 3-bromobenzoylbenzene, 3-bromophenyl phenyl ketone, 3-bromophenyl phenyl methanone, 3-bromophenyl phenylmethanone, 3-bromophenyl-phenyl-methanone, acmc-20an8e, bromobenzophenone,m, m-bromobenzophenone, methanone, 3-bromophenyl phenyl
(3-bromophenyl)-phenylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-808-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents