missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzonitrile 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC106701000

 View more versions of this product

Catalog No. AC106701000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Crystalline Mass, Crystals or Crystalline Powder
White to light yellow or beige
37° to 41°C
C7H4BrN
6952-59-6
BrC6H4CN
98.5
Authentic
182.02
100g
09, 350
100.0
3-Bromobenzonitrile
99%
99%
MFCD00001796
Glass bottle

Chemical Identifiers

6952-59-6
100.0
C7H4BrN
182.02
3-Bromobenzonitrile, 99%
98.5
99%
BrC6H4CN
MFCD00001796
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-127-9

RTECSNumber : DI2459500

TSCA : TSCA