Please login to your online account to display your discounted pricing

3-Bromobenzonitrile 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  106700100

 View more versions of this product

Catalog No. AC106700100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Mass, Crystals or Crystalline Powder
Formula Weight 182.02
Quantity 10g
Color White to light yellow or beige
Beilstein 09, 350
TSCA TSCA
EINECS Number 230-127-9
Melting Point 37° to 41°C
RTECS Number DI2459500
CAS Max % 100.0
Chemical Name or Material 3-Bromobenzonitrile
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H4BrN
Linear Formula BrC6H4CN
MDL Number MFCD00001796
CAS 6952-59-6
Infrared Spectrum Authentic
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Bromobenzonitrile
Name Note 99%
CAS 6952-59-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4BrN
Linear Formula BrC6H4CN
Formula Weight 182.02
MDL Number MFCD00001796
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents