missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzonitrile, 99%

Chemical Identifiers

CAS 6952-59-6
Molecular Formula C7H4BrN
Formula Weight 182.03
MDL Number MFCD00001796
UN Number UN3439
Beilstein 1858942
Synonym 3-Cyanobromobenzene
Chemical Name or Material 3-Bromobenzonitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1331709 SDS Alfa Aesar™
A1331709
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1331718 SDS Alfa Aesar™
A1331718
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1331730 SDS Alfa Aesar™
A1331730
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
3-Bromobenzonitrile is involved in the Suzuki reaction under solvent-free conditions.

Solubility
Soluble in water (0.2 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Description & Specifications

Chemical Identifiers

6952-59-6
182.03
UN3439
3-Cyanobromobenzene
C7H4BrN
MFCD00001796
1858942
3-Bromobenzonitrile

Specifications

6952-59-6
224° to 226°C
113°C (235°F)
182.03
UN3439
3-Cyanobromobenzene
99%
37° to 41°C
10g
C7H4BrN
MFCD00001796
1858942
3-Bromobenzonitrile

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

RTECSNumber : DI2459500