missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzenethiol, TCI America™

$110.00 - $330.00

Chemical Identifiers

CAS 6320-01-0
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00009603
InChI Key HNGQQUDFJDROPY-UHFFFAOYSA-N
Synonym 3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, #
PubChem CID 80597
IUPAC Name 3-bromobenzenethiol
SMILES C1=CC(=CC(=C1)Br)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23485G View Documents TCI America
B23485G
5g Each for $110.00
 
B234825G View Documents TCI America
B234825G
25g Each for $330.00
 
Specifications

Chemical Identifiers

6320-01-0
189.07
HNGQQUDFJDROPY-UHFFFAOYSA-N
80597
C1=CC(=CC(=C1)Br)S
C6H5BrS
MFCD00009603
3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, #
3-bromobenzenethiol

Specifications

121°C
5g
C6H5BrS
1760
HNGQQUDFJDROPY-UHFFFAOYSA-N
3-bromobenzenethiol
80597
Clear Liquid at 20°C
3-Bromobenzenethiol
Yellow
6320-01-0
MFCD00009603
3-bromothiophenol, m-bromobenzenethiol, m-bromothiophenol, 3-bromo thiophenol, 3-bromobenzene-1-thiol, benzenethiol, 3-bromo, 3-bromophenyl mercaptan, 3-bromo-benzenethiol, pubchem6805, #
C1=CC(=CC(=C1)Br)S
189.07
189.07
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator