Please login to your online account to display your discounted pricing

3-Bromobenzenesulfonyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromobenzenesulfonyl chloride
Name Note 98%
CAS 2905-24-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4BrClO2S
Linear Formula BrC6H4SO2Cl
Formula Weight 255.52
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC356580010

  SDS
thermo fisher scientific
AC356580010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 255.52
Physical Form Liquid
Quantity 1g
Color Clear colorless to yellow
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Linear Formula BrC6H4SO2Cl
CAS 2905-24-0
MDL Number MFCD00052313
Chemical Name or Material 3-Bromobenzenesulfonyl chloride
Molecular Formula C6H4BrClO2S
CAS Min % 97.5
Refractive Index 1.5900 to 1.5940 (20°C, 589nm)
Name Note 98%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%