Learn More
3-Bromobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
$119.53 - $1385.66
Chemical Identifiers
CAS | 2905-24-0 |
---|---|
Molecular Formula | C6H4BrClO2S |
Molecular Weight (g/mol) | 255.51 |
MDL Number | MFCD00052313 |
InChI Key | PJGOLCXVWIYXRQ-UHFFFAOYSA-N |
Synonym | 3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo |
PubChem CID | 17943 |
IUPAC Name | 3-bromobenzenesulfonyl chloride |
SMILES | ClS(=O)(=O)C1=CC=CC(Br)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1150703
|
Thermo Scientific Chemicals
A1150703 |
1 g |
Each for $119.53
|
|
|||||
AAA1150706
|
Thermo Scientific Chemicals
A1150706 |
5 g |
Each for $335.82
|
|
|||||
AAA1150714
|
Thermo Scientific Chemicals
A1150714 |
25 g |
Each for $1,385.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2905-24-0 | |
255.51 | |
PJGOLCXVWIYXRQ-UHFFFAOYSA-N | |
17943 | |
ClS(=O)(=O)C1=CC=CC(Br)=C1 |
C6H4BrClO2S | |
MFCD00052313 | |
3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo | |
3-bromobenzenesulfonyl chloride |
Specifications
2905-24-0 | |
1.773 | |
>110°C (230°F) | |
1.593 | |
1 g | |
2691656 | |
3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo | |
ClS(=O)(=O)C1=CC=CC(Br)=C1 | |
255.51 | |
255.52 | |
3-Bromobenzenesulfonyl chloride |
30°C to 33°C | |
90°C to 91°C (0.5 mmHg) | |
C6H4BrClO2S | |
MFCD00052313 | |
UN3261 | |
Moisture sensitive | |
PJGOLCXVWIYXRQ-UHFFFAOYSA-N | |
3-bromobenzenesulfonyl chloride | |
17943 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only