missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1150703

 View more versions of this product

Catalog No. AAA1150703



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromobenzenesulfonyl chloride
30°C to 33°C
90°C to 91°C (0.5 mmHg)
C6H4BrClO2S
MFCD00052313
UN3261
Moisture sensitive
PJGOLCXVWIYXRQ-UHFFFAOYSA-N
3-bromobenzenesulfonyl chloride
17943
97%
2905-24-0
1.773
>110°C (230°F)
1.593
1 g
2691656
3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo
ClS(=O)(=O)C1=CC=CC(Br)=C1
255.51
255.52

Chemical Identifiers

2905-24-0
255.51
PJGOLCXVWIYXRQ-UHFFFAOYSA-N
17943
ClS(=O)(=O)C1=CC=CC(Br)=C1
C6H4BrClO2S
MFCD00052313
3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo
3-bromobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only