missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzenesulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ A1150706

 View more versions of this product

Catalog No. AAA1150706


 

Specifications

Specifications

3-Bromobenzenesulfonyl chloride
97%
MFCD00052313
2691656
90°C to 91°C (0.5mmHg)
>110°C (230°F)
C1=CC(=CC(=C1)Br)S(=O)(=O)Cl
255.51
255.52
2905-24-0
C6H4BrClO2S
UN3261
30°C to 33°C
5g
PJGOLCXVWIYXRQ-UHFFFAOYSA-N
3-bromobenzenesulfonyl chloride
17943

Chemical Identifiers

2905-24-0
255.51
PJGOLCXVWIYXRQ-UHFFFAOYSA-N
C6H4BrClO2S
MFCD00052313
17943
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents