missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzeneboronic acid N-methyldiethanolamine ester, 98%

$58.59 - $188.14

Chemical Identifiers

CAS 374538-00-8
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD02093962
InChI Key CYGPBFLFZQMUBN-UHFFFAOYSA-N
Synonym 2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaborocane, 3-bromobenzeneboronic acid n-methyldiethanolamine ester, 2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaboraperhydroocine, 3-bromophenylboronic acid n-methyl diethanolamine cyclic este
PubChem CID 10039503
IUPAC Name 2-(3-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
SMILES B1(OCCN(CCO1)C)C2=CC(=CC=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1779603 View Documents Alfa Aesar™
L1779603
1g Each for $58.59
 
AAL1779606 View Documents Alfa Aesar™
L1779606
5g Each for $188.14
 
Specifications

Chemical Identifiers

374538-00-8
283.96
CYGPBFLFZQMUBN-UHFFFAOYSA-N
10039503
B1(OCCN(CCO1)C)C2=CC(=CC=C2)Br
C11H15BBrNO2
MFCD02093962
2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaborocane, 3-bromobenzeneboronic acid n-methyldiethanolamine ester, 2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaboraperhydroocine, 3-bromophenylboronic acid n-methyl diethanolamine cyclic este
2-(3-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane

Specifications

128°C to 129°C
374538-00-8
MFCD02093962
CYGPBFLFZQMUBN-UHFFFAOYSA-N
2-(3-bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane
10039503
98%
1g
C11H15BBrNO2
2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaborocane, 3-bromobenzeneboronic acid n-methyldiethanolamine ester, 2-3-bromophenyl-6-methyl-1,3,6,2-dioxazaboraperhydroocine, 3-bromophenylboronic acid n-methyl diethanolamine cyclic este
B1(OCCN(CCO1)C)C2=CC(=CC=C2)Br
283.96
283.96
3-Bromobenzeneboronic acid N-methyldiethanolamine ester
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning