missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromobenzamide, TCI America™

$59.58 - $178.75

Chemical Identifiers

CAS 22726-00-7
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00017127
InChI Key ODJFDWIECLJWSR-UHFFFAOYSA-N
Synonym m-bromobenzamide, benzamide, 3-bromo, 3-bromo-benzamide, benzamide, m-bromo, 3-carbamoyl bromobenzene, 3-bromanylbenzamide, pubchem3734, 3-bromophenylcarboxamide, acmc-209fyc, 3-bromobenzoic acid amide
PubChem CID 89807
IUPAC Name 3-bromobenzamide
SMILES C1=CC(=CC(=C1)Br)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B51785G View Documents TCI America
B51785G
5g Each for $59.58
 
B517825G View Documents TCI America
B517825G
25g Each for $178.75
 
Specifications

Chemical Identifiers

22726-00-7
200.035
ODJFDWIECLJWSR-UHFFFAOYSA-N
89807
C1=CC(=CC(=C1)Br)C(=O)N
C7H6BrNO
MFCD00017127
m-bromobenzamide, benzamide, 3-bromo, 3-bromo-benzamide, benzamide, m-bromo, 3-carbamoyl bromobenzene, 3-bromanylbenzamide, pubchem3734, 3-bromophenylcarboxamide, acmc-209fyc, 3-bromobenzoic acid amide
3-bromobenzamide

Specifications

White-Yellow
5g
C7H6BrNO
m-bromobenzamide, benzamide, 3-bromo, 3-bromo-benzamide, benzamide, m-bromo, 3-carbamoyl bromobenzene, 3-bromanylbenzamide, pubchem3734, 3-bromophenylcarboxamide, acmc-209fyc, 3-bromobenzoic acid amide
C1=CC(=CC(=C1)Br)C(=O)N
200.035
200.04
≥98.0% (HPLC,N)
158°C
22726-00-7
MFCD00017127
ODJFDWIECLJWSR-UHFFFAOYSA-N
3-bromobenzamide
89807
Crystal-Powder at 20°C
3-Bromobenzamide
Safety and Handling

Safety and Handling

TSCA : No