Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzaldoxime, 97%

$29.08 - $274.23

Chemical Identifiers

Chemical Name or Material 3-Bromobenzaldoxime
CAS 51873-95-1
Molecular Formula C7H6BrNO
Formula Weight 200.03
MDL Number MFCD00143098
Beilstein 2040559
Synonym 3-Bromobenzaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2518103

  SDS
alfa aesar™
B2518103
1g Each for $29.08

AAB2518106

  SDS
alfa aesar™
B2518106
5g Each for $78.80

AAB2518114

  SDS
alfa aesar™
B2518114
25g Each for $274.23
Description & Specifications

Specifications

Formula Weight 200.03
Chemical Name or Material 3-Bromobenzaldoxime
Melting Point 70° to 74°C
Quantity 1g
Assay 97%
Molecular Formula C7H6BrNO
MDL Number MFCD00143098
CAS 51873-95-1
Beilstein 2040559
Synonym 3-Bromobenzaldehyde oxime
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.