Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzaldehyde diethyl acetal, 97%

$29.38 - $70.96

Chemical Identifiers

Chemical Name or Material 3-Bromobenzaldehyde diethyl acetal
CAS 75148-49-1
Molecular Formula C11H15BrO2
Formula Weight 259.14
MDL Number MFCD01075697
Beilstein 6392356
Synonym 3-Bromo-alpha,alpha-diethoxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1551406

  SDS
alfa aesar™
L1551406
5g Each for $29.38

AAL1551414

  SDS
alfa aesar™
L1551414
25g Each for $70.96
Description & Specifications

Specifications

Formula Weight 259.14
CAS 75148-49-1
Chemical Name or Material 3-Bromobenzaldehyde diethyl acetal
MDL Number MFCD01075697
Molecular Formula C11H15BrO2
Assay 97%
Synonym 3-Bromo-alpha,alpha-diethoxytoluene
Flash Point 104°C (219°F)
Boiling Point 88° to 91°C (0.8mmHg)
Beilstein 6392356
Quantity 5g