Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromobenzaldehyde diethyl acetal, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1551406

 View more versions of this product

Catalog No. AAL1551406

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 259.14
Chemical Name or Material 3-Bromobenzaldehyde diethyl acetal
MDL Number MFCD01075697
CAS 75148-49-1
Assay 97%
Molecular Formula C11H15BrO2
Beilstein 6392356
Synonym 3-Bromo-alpha,alpha-diethoxytoluene
Boiling Point 88° to 91°C (0.8mmHg)
Quantity 5g
Flash Point 104°C (219°F)

Specifications

Chemical Name or Material 3-Bromobenzaldehyde diethyl acetal
CAS 75148-49-1
Molecular Formula C11H15BrO2
Formula Weight 259.14
MDL Number MFCD01075697
Beilstein 6392356
Synonym 3-Bromo-alpha,alpha-diethoxytoluene