missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromoanisole, 98+%

Manufacturer:  Alfa Aesar™ A1562536

 View more versions of this product

Catalog No. AAA1562536



Description

Description

Applications
3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.

Solubility
Miscible in methanol, benzene, hexane, toluene and dichloromethane. Practically insoluble in water.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

3-Bromoanisole
210°C to 211°C
500g
C7H7BrO
1524687
PLDWAJLZAAHOGG-UHFFFAOYSA-N
1-bromo-3-methoxybenzene
16971
≥98%
93°C (199°F)
~2°C
2398-37-0
MFCD00000081
3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
COC1=CC(=CC=C1)Br
187.036
187.04

Chemical Identifiers

2398-37-0
187.036
PLDWAJLZAAHOGG-UHFFFAOYSA-N
16971
COC1=CC(=CC=C1)Br
C7H7BrO
MFCD00000081
3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole
1-bromo-3-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents