missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 3-Bromoanisole, 98+%
Manufacturer: Alfa Aesar™ A1562514
Description
Applications3-Bromoanisole used in chemical reaction as intermediates to obtain target materials such as dyes, pharmaceuticals, perfumes, photoinitiators and agrochemicals.
Solubility
Miscible in methanol, benzene, hexane, toluene and dichloromethane. Practically insoluble in water.
Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Specifications
3-Bromoanisole | |
210°C to 211°C | |
25g | |
C7H7BrO | |
1524687 | |
PLDWAJLZAAHOGG-UHFFFAOYSA-N | |
1-bromo-3-methoxybenzene | |
16971 | |
≥98% |
93°C (199°F) | |
~2°C | |
2398-37-0 | |
MFCD00000081 | |
3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole | |
COC1=CC(=CC=C1)Br | |
187.036 | |
187.04 |
Chemical Identifiers
2398-37-0 | |
187.036 | |
PLDWAJLZAAHOGG-UHFFFAOYSA-N | |
16971 | |
COC1=CC(=CC=C1)Br |
C7H7BrO | |
MFCD00000081 | |
3-bromoanisole, m-bromoanisole, 3-methoxybromobenzene, 3-bromo anisole, m-bromomethoxybenzene, benzene, 1-bromo-3-methoxy, anisole, m-bromo, 3-methoxy-1-bromobenzene, m-methoxybromobenzene, meta-bromoanisole | |
1-bromo-3-methoxybenzene |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA