Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Bromoacetophenone diethyl ketal, 96%, stab. with Potassium carbonate

$92.99 - $310.93

Chemical Identifiers

Chemical Name or Material 3'-Bromoacetophenone diethyl ketal
CAS 480439-43-8
Name Note stabilized with Potassium carbonate
Molecular Formula C12H17BrO2
Formula Weight 273.17
MDL Number MFCD14636457
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3215706

 
alfa aesar™
H3215706
5g Each for $92.99

AAH3215714

 
alfa aesar™
H3215714
25g Each for $310.93
Description & Specifications

Specifications

Formula Weight 273.17
Chemical Name or Material 3'-Bromoacetophenone diethyl ketal
MDL Number MFCD14636457
Name Note stabilized with Potassium carbonate
Assay 96%
Molecular Formula C12H17BrO2
CAS 480439-43-8
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.