Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Bromoacetophenone, 98+%

$62.96 - $709.61

Chemical Identifiers

Chemical Name or Material 3'-Bromoacetophenone
CAS 2142-63-4
Molecular Formula C8H7BrO
Formula Weight 199.05
MDL Number MFCD00000083
Beilstein 507599
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1245414

  SDS
alfa aesar™
A1245414
25g Each for $62.96

AAA1245422

  SDS
alfa aesar™
A1245422
100g Each for $184.72

AAA1245436

  SDS
alfa aesar™
A1245436
500g Each for $709.61
Description & Specifications

Specifications

Formula Weight 199.05
Chemical Name or Material 3'-Bromoacetophenone
TSCA TSCA
Assay ≥98%
Molecular Formula C8H7BrO
MDL Number MFCD00000083
CAS 2142-63-4
Melting Point 7° to 8°C
Beilstein 507599
Boiling Point 79° to 81°C (2mmHg)
Quantity 25g
Flash Point 114°C (237°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.