missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3'-Bromoacetophenone, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1245422
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3'-Bromoacetophenone | |
7°C to 8°C | |
79°C to 81°C (2 mmHg) | |
C8H7BrO | |
MFCD00000083 | |
507599 | |
JYAQYXOVOHJRCS-UHFFFAOYSA-N | |
1-(3-bromophenyl)ethanone | |
16502 | |
≥98% |
2142-63-4 | |
1.498 | |
114°C (237°F) | |
1.575 | |
100 g | |
3'-bromoacetophenone, m-bromoacetophenone, 1-3-bromophenyl ethanone, 1-3-bromophenyl ethan-1-one, ethanone, 1-3-bromophenyl, 1-acetyl-3-bromobenzene, 3-bromoacetophenone, 1-3-bromo-phenyl-ethanone, 3-bromo acetophenone, acetophenone, 3'-bromo | |
CC(=O)C1=CC(=CC=C1)Br | |
199.047 | |
199.05 |
Chemical Identifiers
2142-63-4 | |
199.047 | |
JYAQYXOVOHJRCS-UHFFFAOYSA-N | |
16502 | |
CC(=O)C1=CC(=CC=C1)Br |
C8H7BrO | |
MFCD00000083 | |
3'-bromoacetophenone, m-bromoacetophenone, 1-3-bromophenyl ethanone, 1-3-bromophenyl ethan-1-one, ethanone, 1-3-bromophenyl, 1-acetyl-3-bromobenzene, 3-bromoacetophenone, 1-3-bromo-phenyl-ethanone, 3-bromo acetophenone, acetophenone, 3'-bromo | |
1-(3-bromophenyl)ethanone |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-396-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only