missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3'-Bromoacetophenone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC106581000

 View more versions of this product

Catalog No. AC106581000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Liquid
Clear slightly yellow to yellow
96.0
2142-63-4
MFCD00000083
1.5749 to 1.5769 (20°C, 589nm)
Authentic
C8H7BrO
199.05
100g
1.4900g/mL
100.0
BrC6H4COCH3
97%
1-(3-Bromophenyl)ethanone
Glass bottle
3'-Bromoacetophenone

Chemical Identifiers

2142-63-4
100.0
C8H7BrO
199.05
1-(3-Bromophenyl)ethanone
≥96.0%
96.0
97%
BrC6H4COCH3
MFCD00000083
3'-Bromoacetophenone

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning