missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-o-xylene, TCI America™

$45.83 - $160.42

Chemical Identifiers

CAS 576-23-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000069
InChI Key WLPXNBYWDDYJTN-UHFFFAOYSA-N
Synonym 3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
PubChem CID 68472
IUPAC Name 1-bromo-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)Br)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12825G View Documents TCI America
B12825G
5g Each for $45.83
 
B128225G View Documents TCI America
B128225G
25g Each for $160.42
 
Specifications

Chemical Identifiers

576-23-8
185.064
WLPXNBYWDDYJTN-UHFFFAOYSA-N
68472
CC1=C(C(=CC=C1)Br)C
C8H9Br
MFCD00000069
3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
1-bromo-2,3-dimethylbenzene

Specifications

214°C
5g
C8H9Br
3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
CC1=C(C(=CC=C1)Br)C
185.064
185.06
≥98.0% (GC)
Yellow
576-23-8
MFCD00000069
WLPXNBYWDDYJTN-UHFFFAOYSA-N
1-bromo-2,3-dimethylbenzene
68472
Clear Liquid at 20°C
3-Bromo-o-xylene
Safety and Handling

Safety and Handling

TSCA : No