Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-o-xylene, 99%

$61.64 - $858.85

Chemical Identifiers

Chemical Name or Material 3-Bromo-o-xylene
CAS 576-23-8
Molecular Formula C8H9Br
Formula Weight 185.07
MDL Number MFCD00000069
Beilstein 2077312
Synonym 3-Bromo-1,2-dimethylbenzene; 2,3-Dimethylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1588906

  SDS
alfa aesar™
A1588906
5g Each for $61.64

AAA1588922

  SDS
alfa aesar™
A1588922
100g Each for $858.85
Description & Specifications

Specifications

Formula Weight 185.07
Synonym 3-Bromo-1,2-dimethylbenzene; 2,3-Dimethylbromobenzene
Flash Point 80°C (176°F)
Beilstein 2077312
Assay 99%
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 212° to 214°C
Quantity 5g
MDL Number MFCD00000069
Chemical Name or Material 3-Bromo-o-xylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Molecular Formula C8H9Br
CAS 576-23-8

Applications
3-Bromo-o-xylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.