Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-N-methylaniline, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromo-N-methylaniline
CAS 66584-32-5
Formula Weight 186.05
MDL Number MFCD05664376
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5546403

  SDS
alfa aesar™
H5546403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5546406

  SDS
alfa aesar™
H5546406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5546414

  SDS
alfa aesar™
H5546414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.05
Chemical Name or Material 3-Bromo-N-methylaniline
Density 1.461g/mL
Boiling Point 253°C
Quantity 1g
Odor Unpleasant
Color Colorless to yellow
Physical Form Liquid
Assay 97%
MDL Number MFCD05664376
CAS 66584-32-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.6210
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.