missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-N-ethylbenzenesulfonamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 871269-07-7
Molecular Formula C8H10BrNO2S
Molecular Weight (g/mol) 264.137
MDL Number MFCD07363819
InChI Key MADIDEQQKAOBFI-UHFFFAOYSA-N
Synonym 3-bromo-n-ethylbenzenesulphonamide, n-ethyl 3-bromobenzenesulfonamide, 3-bromo-n-ethylbenzene-1-sulfonamide, benzenesulfonamide, 3-bromo-n-ethyl, acmc-209qfl, 3-bromo-n-ethylbenzenesulfomide, n-ethyl-3-bromobenzenesulfonamide, benzenesulfonamide,3-bromo-n-ethyl, 3-bromanyl-n-ethyl-benzenesulfonamide, n-ethyl 3-bromophenylsulfonamide
PubChem CID 7213268
IUPAC Name 3-bromo-N-ethylbenzenesulfonamide
SMILES CCNS(=O)(=O)C1=CC(=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60276MD View Documents Alfa Aesar™
H60276MD
250mg Each for $220.53
 
AAH6027603 View Documents Alfa Aesar™
H6027603
1g Each for $703.20
 
Specifications

Chemical Identifiers

871269-07-7
264.137
MADIDEQQKAOBFI-UHFFFAOYSA-N
7213268
CCNS(=O)(=O)C1=CC(=CC=C1)Br
C8H10BrNO2S
MFCD07363819
3-bromo-n-ethylbenzenesulphonamide, n-ethyl 3-bromobenzenesulfonamide, 3-bromo-n-ethylbenzene-1-sulfonamide, benzenesulfonamide, 3-bromo-n-ethyl, acmc-209qfl, 3-bromo-n-ethylbenzenesulfomide, n-ethyl-3-bromobenzenesulfonamide, benzenesulfonamide,3-bromo-n-ethyl, 3-bromanyl-n-ethyl-benzenesulfonamide, n-ethyl 3-bromophenylsulfonamide
3-bromo-N-ethylbenzenesulfonamide

Specifications

46°C to 50°C
871269-07-7
MFCD07363819
MADIDEQQKAOBFI-UHFFFAOYSA-N
3-bromo-N-ethylbenzenesulfonamide
7213268
97%
250mg
C8H10BrNO2S
3-bromo-n-ethylbenzenesulphonamide, n-ethyl 3-bromobenzenesulfonamide, 3-bromo-n-ethylbenzene-1-sulfonamide, benzenesulfonamide, 3-bromo-n-ethyl, acmc-209qfl, 3-bromo-n-ethylbenzenesulfomide, n-ethyl-3-bromobenzenesulfonamide, benzenesulfonamide,3-bromo-n-ethyl, 3-bromanyl-n-ethyl-benzenesulfonamide, n-ethyl 3-bromophenylsulfonamide
CCNS(=O)(=O)C1=CC(=CC=C1)Br
264.137
264.14
3-Bromo-N-ethylbenzenesulfonamide