missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-N-(3-pyridylmethyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 849136-14-7
Molecular Formula C12H11BrN2O2S
Molecular Weight (g/mol) 327.196
MDL Number MFCD07356489
InChI Key VAQOTDUYHINTAB-UHFFFAOYSA-N
Synonym 3-bromo-n-pyridin-3-ylmethyl benzenesulfonamide, 3-bromo-n-3-pyridylmethyl benzenesulfonamide, 3-bromo-n-pyridin-3-yl methyl benzene-1-sulfonamide
PubChem CID 3772600
IUPAC Name 3-bromo-N-(pyridin-3-ylmethyl)benzenesulfonamide
SMILES C1=CC(=CC(=C1)Br)S(=O)(=O)NCC2=CN=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60958MD View Documents Alfa Aesar™
H60958MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6095803 View Documents Alfa Aesar™
H6095803
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

849136-14-7
327.196
VAQOTDUYHINTAB-UHFFFAOYSA-N
3772600
C1=CC(=CC(=C1)Br)S(=O)(=O)NCC2=CN=CC=C2
C12H11BrN2O2S
MFCD07356489
3-bromo-n-pyridin-3-ylmethyl benzenesulfonamide, 3-bromo-n-3-pyridylmethyl benzenesulfonamide, 3-bromo-n-pyridin-3-yl methyl benzene-1-sulfonamide
3-bromo-N-(pyridin-3-ylmethyl)benzenesulfonamide

Specifications

250mg
C12H11BrN2O2S
3-bromo-n-pyridin-3-ylmethyl benzenesulfonamide, 3-bromo-n-3-pyridylmethyl benzenesulfonamide, 3-bromo-n-pyridin-3-yl methyl benzene-1-sulfonamide
C1=CC(=CC(=C1)Br)S(=O)(=O)NCC2=CN=CC=C2
327.196
327.2
3-Bromo-N-(3-pyridylmethyl)benzenesulfonamide
849136-14-7
MFCD07356489
VAQOTDUYHINTAB-UHFFFAOYSA-N
3-bromo-N-(pyridin-3-ylmethyl)benzenesulfonamide
3772600
97%