missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-N-(2,3-dimethylphenyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 849228-34-8
Molecular Formula C14H14BrNO2S
Molecular Weight (g/mol) 340.235
MDL Number MFCD07708351
InChI Key GAZGMCMRCZAATD-UHFFFAOYSA-N
Synonym 3-bromo-n-2,3-dimethylphenyl benzenesulfonamide, 3-bromo-n-2,3-dimethyl-phenyl-benzenesulfonamide
PubChem CID 4816670
IUPAC Name 3-bromo-N-(2,3-dimethylphenyl)benzenesulfonamide
SMILES CC1=C(C(=CC=C1)NS(=O)(=O)C2=CC(=CC=C2)Br)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60002MD View Documents Alfa Aesar™
H60002MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6000203 View Documents Alfa Aesar™
H6000203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

849228-34-8
340.235
GAZGMCMRCZAATD-UHFFFAOYSA-N
4816670
CC1=C(C(=CC=C1)NS(=O)(=O)C2=CC(=CC=C2)Br)C
C14H14BrNO2S
MFCD07708351
3-bromo-n-2,3-dimethylphenyl benzenesulfonamide, 3-bromo-n-2,3-dimethyl-phenyl-benzenesulfonamide
3-bromo-N-(2,3-dimethylphenyl)benzenesulfonamide

Specifications

250mg
C14H14BrNO2S
3-bromo-n-2,3-dimethylphenyl benzenesulfonamide, 3-bromo-n-2,3-dimethyl-phenyl-benzenesulfonamide
CC1=C(C(=CC=C1)NS(=O)(=O)C2=CC(=CC=C2)Br)C
340.235
340.23
3-Bromo-N-(2,3-dimethylphenyl)benzenesulfonamide
849228-34-8
MFCD07708351
GAZGMCMRCZAATD-UHFFFAOYSA-N
3-bromo-N-(2,3-dimethylphenyl)benzenesulfonamide
4816670
97%