missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-DL-phenylalanine, 95%

$106.72 - $379.36

Chemical Identifiers

CAS 30163-20-3
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD05227935
InChI Key GDMOHOYNMWWBAU-UHFFFAOYSA-N
Synonym 2-amino-3-3-bromophenyl propanoic acid, 3-bromo-dl-phenylalanine, 3-bromophenylalanine, dl-3-br-phe-oh, h-dl-phe 3-br-oh, dl-3-bromophenylalanine, pubchem18643, 3-bromophenylalanine #, 2-3-bromobenzyl glycine, dl-3-br-phe
PubChem CID 554078
IUPAC Name 2-amino-3-(3-bromophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)Br)CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6554703 View Documents Alfa Aesar™
H6554703
1g Each for $106.72
 
AAH6554706 View Documents Alfa Aesar™
H6554706
5g Each for $379.36
 
Specifications

Chemical Identifiers

30163-20-3
244.088
GDMOHOYNMWWBAU-UHFFFAOYSA-N
554078
C1=CC(=CC(=C1)Br)CC(C(=O)O)N
C9H10BrNO2
MFCD05227935
2-amino-3-3-bromophenyl propanoic acid, 3-bromo-dl-phenylalanine, 3-bromophenylalanine, dl-3-br-phe-oh, h-dl-phe 3-br-oh, dl-3-bromophenylalanine, pubchem18643, 3-bromophenylalanine #, 2-3-bromobenzyl glycine, dl-3-br-phe
2-amino-3-(3-bromophenyl)propanoic acid

Specifications

30163-20-3
MFCD05227935
2-amino-3-3-bromophenyl propanoic acid, 3-bromo-dl-phenylalanine, 3-bromophenylalanine, dl-3-br-phe-oh, h-dl-phe 3-br-oh, dl-3-bromophenylalanine, pubchem18643, 3-bromophenylalanine #, 2-3-bromobenzyl glycine, dl-3-br-phe
C1=CC(=CC(=C1)Br)CC(C(=O)O)N
244.088
244.09
95%
3-Bromo-DL-phenylalanine
C9H10BrNO2
2939808
GDMOHOYNMWWBAU-UHFFFAOYSA-N
2-amino-3-(3-bromophenyl)propanoic acid
554078
Solid
1g