Learn More
3-Bromo-9-phenylcarbazole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6421214
Description
Used in manufacturing of OLED materials. Also used as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in manufacturing of OLED materials. Also used as intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
3-Bromo-9-phenylcarbazole | |
97°C to 98°C | |
MFCD11977305 | |
1315492 | |
Slightly soluble in water. | |
BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 | |
322.21 | |
322.2 | |
Solid |
1153-85-1 | |
C18H12BrN | |
25 g | |
3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole | |
KUBSCXXKQGDPPD-UHFFFAOYSA-N | |
3-bromo-9-phenylcarbazole | |
18942624 | |
98% |
Chemical Identifiers
1153-85-1 | |
322.21 | |
KUBSCXXKQGDPPD-UHFFFAOYSA-N | |
18942624 | |
BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 |
C18H12BrN | |
MFCD11977305 | |
3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole | |
3-bromo-9-phenylcarbazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only