missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-9-(4-bromophenyl)-9H-carbazole, TCI America™

$265.84 - $922.80

Chemical Identifiers

CAS 1226860-66-7
Molecular Formula C18H11Br2N
Molecular Weight (g/mol) 401.101
InChI Key ZUNLPNWEQKBULG-UHFFFAOYSA-N
PubChem CID 66797808
IUPAC Name 3-bromo-9-(4-bromophenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2C4=CC=C(C=C4)Br)C=CC(=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B51075G View Documents TCI America
B51075G
5g Each for $922.80
 
B51071G View Documents TCI America
B51071G
1g Each for $265.84
 
Specifications

Chemical Identifiers

1226860-66-7
401.101
66797808
C1=CC=C2C(=C1)C3=C(N2C4=CC=C(C=C4)Br)C=CC(=C3)Br
C18H11Br2N
ZUNLPNWEQKBULG-UHFFFAOYSA-N
3-bromo-9-(4-bromophenyl)carbazole

Specifications

White
5g
C18H11Br2N
C1=CC=C2C(=C1)C3=C(N2C4=CC=C(C=C4)Br)C=CC(=C3)Br
401.101
401.10
≥97.0% (GC)
150°C
1226860-66-7
ZUNLPNWEQKBULG-UHFFFAOYSA-N
3-bromo-9-(4-bromophenyl)carbazole
66797808
Crystal-Powder at 20°C
3-Bromo-9-(4-bromophenyl)-9H-carbazole
Safety and Handling

Safety and Handling

TSCA : No