missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromo-9-(1-naphthyl)-9H-carbazole, TCI America™

$128.34 - $383.48

Chemical Identifiers

CAS 934545-83-2
Molecular Formula C22H14BrN
Molecular Weight (g/mol) 372.265
MDL Number MFCD26127423
InChI Key QTWVKYHIDBPQIL-UHFFFAOYSA-N
PubChem CID 59561149
IUPAC Name 3-bromo-9-naphthalen-1-ylcarbazole
SMILES C1=CC=C2C(=C1)C=CC=C2N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B50501G View Documents TCI America
B50501G
1g Each for $383.48
 
B5050200MG View Documents TCI America
B5050200MG
200mg Each for $128.34
 
Specifications

Chemical Identifiers

934545-83-2
372.265
QTWVKYHIDBPQIL-UHFFFAOYSA-N
3-bromo-9-naphthalen-1-ylcarbazole
C22H14BrN
MFCD26127423
59561149
C1=CC=C2C(=C1)C=CC=C2N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53

Specifications

White-Yellow
934545-83-2
MFCD26127423
C1=CC=C2C(=C1)C=CC=C2N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53
372.265
372.27
≥97.0% (GC)
1g
C22H14BrN
QTWVKYHIDBPQIL-UHFFFAOYSA-N
3-bromo-9-naphthalen-1-ylcarbazole
59561149
Crystal-Powder at 20°C
3-Bromo-9-(1-naphthyl)-9H-carbazole
Safety and Handling

Safety and Handling

TSCA : No