missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-7-nitroindole, 97%

$23.14 - $225.15

Chemical Identifiers

CAS 397864-11-8
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.044
MDL Number MFCD02684155
InChI Key ZJCDMQUXOJHHBJ-UHFFFAOYSA-N
Synonym 3-bromo-7-nitroindole, 1h-indole, 3-bromo-7-nitro, 1h-indole,3-bromo-7-nitro, 6-methoxy-quinolin-3-ylamine
PubChem CID 17750599
IUPAC Name 3-bromo-7-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63940MD View Documents Alfa Aesar™
H63940MD
250mg Each for $23.14
 
AAH6394003 View Documents Alfa Aesar™
H6394003
1g Each for $60.77
 
AAH6394006 View Documents Alfa Aesar™
H6394006
5g Each for $225.15
 
Specifications

Chemical Identifiers

397864-11-8
241.044
ZJCDMQUXOJHHBJ-UHFFFAOYSA-N
17750599
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2Br
C8H5BrN2O2
MFCD02684155
3-bromo-7-nitroindole, 1h-indole, 3-bromo-7-nitro, 1h-indole,3-bromo-7-nitro, 6-methoxy-quinolin-3-ylamine
3-bromo-7-nitro-1H-indole

Specifications

250mg
C8H5BrN2O2
3-bromo-7-nitroindole, 1h-indole, 3-bromo-7-nitro, 1h-indole,3-bromo-7-nitro, 6-methoxy-quinolin-3-ylamine
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2Br
241.044
241.04
97%
397864-11-8
MFCD02684155
ZJCDMQUXOJHHBJ-UHFFFAOYSA-N
3-bromo-7-nitro-1H-indole
17750599
Solid
3-Bromo-7-nitroindole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C