missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-6-methoxy-2-methylpyridine, 98%

Manufacturer:  Alfa Aesar™ H6196206

 View more versions of this product

Catalog No. AAH6196206



Specifications

Specifications

3-Bromo-6-methoxy-2-methylpyridine
5g
C7H8BrNO
5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
CC1=C(C=CC(=N1)OC)Br
202.051
202.05
98%
Colorless
126717-59-7
MFCD03094943
VWNXCCTWVAQAPL-UHFFFAOYSA-N
3-bromo-6-methoxy-2-methylpyridine
10798146
Liquid

Chemical Identifiers

126717-59-7
202.051
VWNXCCTWVAQAPL-UHFFFAOYSA-N
10798146
CC1=C(C=CC(=N1)OC)Br
C7H8BrNO
MFCD03094943
5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
3-bromo-6-methoxy-2-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents