missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-6,9-diphenylcarbazole 98.0+%, TCI America™
$66.37 - $235.57
Chemical Identifiers
CAS | 1160294-85-8 |
---|---|
Molecular Formula | C24H16BrN |
Molecular Weight (g/mol) | 398.30 |
MDL Number | MFCD20257783 |
InChI Key | JBEQRROTSDUCGC-UHFFFAOYSA-N |
PubChem CID | 57746295 |
IUPAC Name | 3-bromo-6,9-diphenyl-9H-carbazole |
SMILES | BrC1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
1160294-85-8 | |
398.30 | |
JBEQRROTSDUCGC-UHFFFAOYSA-N | |
3-bromo-6,9-diphenyl-9H-carbazole |
C24H16BrN | |
MFCD20257783 | |
57746295 | |
BrC1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
1160294-85-8 | |
White-Yellow | |
MFCD20257783 | |
JBEQRROTSDUCGC-UHFFFAOYSA-N | |
3-bromo-6,9-diphenyl-9H-carbazole | |
57746295 | |
≥98.0% (GC) | |
3-Bromo-6,9-diphenylcarbazole |
145°C | |
C24H16BrN | |
1 g | |
BrC1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1 | |
398.30 | |
398.30 | |
Crystalline Powder |
Safety and Handling
TSCA : No