missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-5-(trifluoromethyl)pyridine 98.0+%, TCI America™
$290.83 - $867.78
Chemical Identifiers
CAS | 436799-33-6 |
---|---|
Molecular Formula | C6H3BrF3N |
Molecular Weight (g/mol) | 225.996 |
MDL Number | MFCD04972700 |
InChI Key | HEDHNDVPKRVQPN-UHFFFAOYSA-N |
Synonym | 3-bromo-5-trifluormethyl-pyridine, 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 3-bromo-5-trifluoromethylpyridin, 3-trifluoromethyl-5-bromopyridine, 5-bromo-3-trifluoromethyl pyridine, abbypharma ap-11-5533, acmc-209jv5, pubchem3010, pyridine, 3-bromo-5-trifluoromethyl |
PubChem CID | 11127991 |
IUPAC Name | 3-bromo-5-(trifluoromethyl)pyridine |
SMILES | C1=C(C=NC=C1Br)C(F)(F)F |
Chemical Identifiers
436799-33-6 | |
225.996 | |
HEDHNDVPKRVQPN-UHFFFAOYSA-N | |
11127991 | |
C1=C(C=NC=C1Br)C(F)(F)F |
C6H3BrF3N | |
MFCD04972700 | |
3-bromo-5-trifluormethyl-pyridine, 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 3-bromo-5-trifluoromethylpyridin, 3-trifluoromethyl-5-bromopyridine, 5-bromo-3-trifluoromethyl pyridine, abbypharma ap-11-5533, acmc-209jv5, pubchem3010, pyridine, 3-bromo-5-trifluoromethyl | |
3-bromo-5-(trifluoromethyl)pyridine |
Specifications
66°C | |
44°C | |
436799-33-6 | |
MFCD04972700 | |
3-bromo-5-trifluormethyl-pyridine, 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 3-bromo-5-trifluoromethylpyridin, 3-trifluoromethyl-5-bromopyridine, 5-bromo-3-trifluoromethyl pyridine, abbypharma ap-11-5533, acmc-209jv5, pubchem3010, pyridine, 3-bromo-5-trifluoromethyl | |
C1=C(C=NC=C1Br)C(F)(F)F | |
225.996 | |
226.00 | |
≥98.0% (GC,T) |
White-Yellow | |
1g | |
C6H3BrF3N | |
2811 | |
HEDHNDVPKRVQPN-UHFFFAOYSA-N | |
3-bromo-5-(trifluoromethyl)pyridine | |
11127991 | |
Crystal-Lump at 20°C | |
3-Bromo-5-(trifluoromethyl)pyridine |
Safety and Handling
TSCA : No