missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Bromo-5-(trifluoromethyl)pyridine, 97%, ACROS Organics™
Chemical Identifiers
CAS | 436799-33-6 |
---|---|
Molecular Formula | C6H3BrF3N |
Molecular Weight (g/mol) | 226 |
MDL Number | MFCD04972700 |
InChI Key | HEDHNDVPKRVQPN-UHFFFAOYSA-N |
Synonym | 3-bromo-5-trifluormethyl-pyridine, 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 3-bromo-5-trifluoromethylpyridin, 3-trifluoromethyl-5-bromopyridine, 5-bromo-3-trifluoromethyl pyridine, abbypharma ap-11-5533, acmc-209jv5, pubchem3010, pyridine, 3-bromo-5-trifluoromethyl |
PubChem CID | 11127991 |
IUPAC Name | 3-bromo-5-(trifluoromethyl)pyridine |
SMILES | C1=C(C=NC=C1Br)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC429890010
|
Thermo Fisher Scientific
AC429890010 |
1g | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC429890050
|
Thermo Fisher Scientific
AC429890050 |
5g | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
436799-33-6 | |
226 | |
HEDHNDVPKRVQPN-UHFFFAOYSA-N | |
11127991 | |
C1=C(C=NC=C1Br)C(F)(F)F |
C6H3BrF3N | |
MFCD04972700 | |
3-bromo-5-trifluormethyl-pyridine, 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 3-bromo-5-trifluoromethylpyridin, 3-trifluoromethyl-5-bromopyridine, 5-bromo-3-trifluoromethyl pyridine, abbypharma ap-11-5533, acmc-209jv5, pubchem3010, pyridine, 3-bromo-5-trifluoromethyl | |
3-bromo-5-(trifluoromethyl)pyridine |
Specifications
85°C | |
Glass Bottle | |
White to Yellow | |
1g | |
96.0 | |
96% min. (GC) | |
MFCD04972700 | |
HEDHNDVPKRVQPN-UHFFFAOYSA-N | |
3-bromo-5-(trifluoromethyl)pyridine | |
11127991 | |
Crystalline Powder or Solid | |
3-Bromo-5-(trifluoromethyl)pyridine |
Authentic | |
65.0°C to 66.0°C (19.0mmHg) | |
43.5°C to 46.0°C | |
436799-33-6 | |
100.0 | |
C6H3BrF3N | |
3-bromo-5-trifluormethyl-pyridine, 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 3-bromo-5-trifluoromethylpyridin, 3-trifluoromethyl-5-bromopyridine, 5-bromo-3-trifluoromethyl pyridine, abbypharma ap-11-5533, acmc-209jv5, pubchem3010, pyridine, 3-bromo-5-trifluoromethyl | |
C1=C(C=NC=C1Br)C(F)(F)F | |
226 | |
226 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning