Please login to your online account to display your discounted pricing

3-Bromo-5-(trifluoromethyl)pyridine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC429890050

 View more versions of this product

Catalog No. AC429890050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Physical Form Crystalline Powder or Solid
Melting Point 43° to 47°C
Formula Weight 226
Quantity 5g
Color White to yellow
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)pyridine
MDL Number MFCD04972700
CAS 436799-33-6
Molecular Formula C6H3BrF3N
Assay Percent Range 97%
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 3-Bromo-5-(trifluoromethyl)pyridine
CAS 436799-33-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C6H3BrF3N
Formula Weight 226
MDL Number MFCD04972700
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.

SDS & Safety Documents