Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-trifluoromethyl-o-phenylenediamine, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-trifluoromethyl-o-phenylenediamine
CAS 113170-72-2
Molecular Formula C7H6BrF3N2
Formula Weight 255.04
MDL Number MFCD00042484
UN Number UN2811
Synonym 3-Bromo-4,5-diaminobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2742903

  SDS
alfa aesar™
H2742903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2742906

  SDS
alfa aesar™
H2742906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 255.04
Assay 97%
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 113170-72-2
Chemical Name or Material 3-Bromo-5-trifluoromethyl-o-phenylenediamine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Synonym 3-Bromo-4,5-diaminobenzotrifluoride
Quantity 1g
UN Number UN2811
Melting Point 61° to 65°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
MDL Number MFCD00042484
Molecular Formula C7H6BrF3N2
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.