missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)benzoic acid, 97%

$50.58 - $664.65

Chemical Identifiers

CAS 328-67-6
Molecular Formula C8H4BrF3O2
Molecular Weight (g/mol) 269.017
MDL Number MFCD03412186
InChI Key AMZBKZQMAZWIJM-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethyl-benzoic acid, 5-bromo-3-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethylbenzoicacid, 3-bromo-5-trifluoromethyl benzoic acid, benzoic acid, 3-bromo-5-trifluoromethyl, pubchem4737, 3-bromo-5-trifluoromethyl benzoicacid, acmc-1clgk, ksc222e0t
PubChem CID 11086788
IUPAC Name 3-bromo-5-(trifluoromethyl)benzoic acid
SMILES C1=C(C=C(C=C1C(F)(F)F)Br)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6377606 View Documents Alfa Aesar™
H6377606
5g Each for $50.58
 
AAH6377614 View Documents Alfa Aesar™
H6377614
25g Each for $200.47
 
AAH6377622 View Documents Alfa Aesar™
H6377622
100g Each for $664.65
 
Specifications

Chemical Identifiers

328-67-6
269.017
AMZBKZQMAZWIJM-UHFFFAOYSA-N
11086788
C1=C(C=C(C=C1C(F)(F)F)Br)C(=O)O
C8H4BrF3O2
MFCD03412186
3-bromo-5-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethyl-benzoic acid, 5-bromo-3-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethylbenzoicacid, 3-bromo-5-trifluoromethyl benzoic acid, benzoic acid, 3-bromo-5-trifluoromethyl, pubchem4737, 3-bromo-5-trifluoromethyl benzoicacid, acmc-1clgk, ksc222e0t
3-bromo-5-(trifluoromethyl)benzoic acid

Specifications

5g
C8H4BrF3O2
3-bromo-5-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethyl-benzoic acid, 5-bromo-3-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethylbenzoicacid, 3-bromo-5-trifluoromethyl benzoic acid, benzoic acid, 3-bromo-5-trifluoromethyl, pubchem4737, 3-bromo-5-trifluoromethyl benzoicacid, acmc-1clgk, ksc222e0t
C1=C(C=C(C=C1C(F)(F)F)Br)C(=O)O
269.017
269.02
97%
328-67-6
MFCD03412186
AMZBKZQMAZWIJM-UHFFFAOYSA-N
3-bromo-5-(trifluoromethyl)benzoic acid
11086788
Solid
3-Bromo-5-(trifluoromethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning