missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)benzoic acid, 97%

Manufacturer:  Alfa Aesar™ H6377622

 View more versions of this product

Catalog No. AAH6377622



Specifications

Specifications

3-Bromo-5-(trifluoromethyl)benzoic acid
328-67-6
MFCD03412186
AMZBKZQMAZWIJM-UHFFFAOYSA-N
3-bromo-5-(trifluoromethyl)benzoic acid
11086788
Solid
100g
C8H4BrF3O2
3-bromo-5-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethyl-benzoic acid, 5-bromo-3-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethylbenzoicacid, 3-bromo-5-trifluoromethyl benzoic acid, benzoic acid, 3-bromo-5-trifluoromethyl, pubchem4737, 3-bromo-5-trifluoromethyl benzoicacid, acmc-1clgk, ksc222e0t
C1=C(C=C(C=C1C(F)(F)F)Br)C(=O)O
269.017
269.02
97%

Chemical Identifiers

328-67-6
269.017
AMZBKZQMAZWIJM-UHFFFAOYSA-N
11086788
C1=C(C=C(C=C1C(F)(F)F)Br)C(=O)O
C8H4BrF3O2
MFCD03412186
3-bromo-5-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethyl-benzoic acid, 5-bromo-3-trifluoromethyl benzoic acid, 3-bromo-5-trifluoromethylbenzoicacid, 3-bromo-5-trifluoromethyl benzoic acid, benzoic acid, 3-bromo-5-trifluoromethyl, pubchem4737, 3-bromo-5-trifluoromethyl benzoicacid, acmc-1clgk, ksc222e0t
3-bromo-5-(trifluoromethyl)benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents