Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 97%

$102.36 - $326.32

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzeneboronic acid pinacol ester
CAS 479411-92-2
Molecular Formula C13H15BBrF3O2
Formula Weight 350.97
MDL Number MFCD12405333
Synonym 2-[3-Bromo-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5171303

 
alfa aesar™
H5171303
1g Each for $326.32

AAH51713MD

 
alfa aesar™
H51713MD
250mg Each for $102.36
Description & Specifications

Specifications

Formula Weight 350.97
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C13H15BBrF3O2
MDL Number MFCD12405333
CAS 479411-92-2
Melting Point 50° to 52°C
Synonym 2-[3-Bromo-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Quantity 1g

Applications
Used as a active pharmaceutical ingredient.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.