Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)aniline, 98%

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-(trifluoromethyl)aniline
CAS 54962-75-3
Molecular Formula C7H5BrF3N
Formula Weight 240.03
MDL Number MFCD00236205
UN Number UN2810
Synonym 3-Amino-5-bromobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2043703

  SDS
alfa aesar™
B2043703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2043706

  SDS
alfa aesar
B2043706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.03
Boiling Point 221° to 223°C
MDL Number MFCD00236205
Quantity 1g
Molecular Formula C7H5BrF3N
Assay 98%
UN Number UN2810
Flash Point 107°C (225°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)aniline
Synonym 3-Amino-5-bromobenzotrifluoride
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
CAS 54962-75-3
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.