Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(trifluoromethoxy)benzoic acid, 97%

$84.23 - $84.23

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzoic acid
CAS 453565-90-7
Molecular Formula C8H4BrF3O3
Formula Weight 285.02
MDL Number MFCD04116041
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2650003

  SDS
alfa aesar™
H2650003
1g Each for $84.23
Description & Specifications

Specifications

Formula Weight 285.02
Molecular Formula C8H4BrF3O3
Melting Point 87° to 89°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD04116041
CAS 453565-90-7
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzoic acid
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.