Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-(trifluoromethoxy)benzamide, 97%

$183.82 - $756.39

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzamide
CAS 914636-32-1
Molecular Formula C8H5BrF3NO2
Formula Weight 284.03
MDL Number MFCD08458075
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3280503

  SDS
alfa aesar™
H3280503
1g Each for $183.82

AAH3280506

  SDS
alfa aesar™
H3280506
5g Each for $756.39
Description & Specifications

Specifications

Formula Weight 284.03
Melting Point 144° to 146°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)benzamide
Assay 97%
Molecular Formula C8H5BrF3NO2
MDL Number MFCD08458075
CAS 914636-32-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.