Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

3-Bromo-5-phenylpyridine, TCI America™

$131.39 - $388.06

Chemical Identifiers

CAS 142137-17-5
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD04114223
InChI Key AWCQJXPOCRXHNK-UHFFFAOYSA-N
PubChem CID 2762904
IUPAC Name 3-bromo-5-phenylpyridine
SMILES C1=CC=C(C=C1)C2=CC(=CN=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
B5151200MG missing translation for 'viewDocuments' TCI America
B5151200MG
200mg Each for $131.39
 
B51511G missing translation for 'viewDocuments' TCI America
B51511G
1g Each for $388.06
 
Specifications

Chemical Identifiers

142137-17-5
234.096
AWCQJXPOCRXHNK-UHFFFAOYSA-N
3-bromo-5-phenylpyridine
C11H8BrN
MFCD04114223
2762904
C1=CC=C(C=C1)C2=CC(=CN=C2)Br

Specifications

101°C
48°C
142137-17-5
MFCD04114223
C1=CC=C(C=C1)C2=CC(=CN=C2)Br
234.096
234.10
≥98.0% (GC,T)
White
200mg
C11H8BrN
AWCQJXPOCRXHNK-UHFFFAOYSA-N
3-bromo-5-phenylpyridine
2762904
Crystal-Powder at 20°C
3-Bromo-5-phenylpyridine
Safety and Handling

Safety and Handling

TSCA : No