Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-nitrobenzotrifluoride, 97%

$125.20 - $375.19

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-nitrobenzotrifluoride
CAS 630125-49-4
Molecular Formula C7H3BrF3NO2
Formula Weight 270.01
MDL Number MFCD03788719
Synonym 1-Bromo-3-nitro-5-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2617103

  SDS
alfa aesar™
H2617103
1g Each for $125.20

AAH2617106

  SDS
alfa aesar™
H2617106
5g Each for $375.19
Description & Specifications

Specifications

Formula Weight 270.01
Molecular Formula C7H3BrF3NO2
MDL Number MFCD03788719
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P337+P313
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
CAS 630125-49-4
Synonym 1-Bromo-3-nitro-5-(trifluoromethyl)benzene
Assay 97%
Chemical Name or Material 3-Bromo-5-nitrobenzotrifluoride
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338-P337+P313
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.