Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-nitrobenzoic acid, 99%

$80.76 - $213.96

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-nitrobenzoic acid
CAS 6307-83-1
Molecular Formula C7H4BrNO4
Formula Weight 246.02
MDL Number MFCD00100098
Beilstein 2051365
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1963303

  SDS
alfa aesar™
L1963303
1g Each for $213.96

AAL19633MD

  SDS
alfa aesar™
L19633MD
250mg Each for $80.76
Description & Specifications

Specifications

Formula Weight 246.02
Chemical Name or Material 3-Bromo-5-nitrobenzoic acid
Assay 99%
Beilstein 2051365
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H4BrNO4
MDL Number MFCD00100098
CAS 6307-83-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 159° to 161°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.