Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-nitro-1H-indazole, 95%

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-nitro-1H-indazole
CAS 67400-25-3
Formula Weight 242.03
MDL Number MFCD07367964
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3797903

 
alfa aesar™
H3797903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3797909

 
alfa aesar™
H3797909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 227° to 229°C
Formula Weight 242.03
Chemical Name or Material 3-Bromo-5-nitro-1H-indazole
Color Pale brown
Physical Form Powder
Quantity 1g
Assay 95%
MDL Number MFCD07367964
CAS 67400-25-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.