Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-methylpyridine, 97%

$129.92 - $427.92

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-methylpyridine
CAS 3430-16-8
Molecular Formula C6H6BrN
Formula Weight 172.03
MDL Number MFCD01646141
Synonym 5-Bromo-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3013703

 
alfa aesar™
H3013703
1g Each for $129.92

AAH3013706

 
alfa aesar™
H3013706
5g Each for $427.92
Description & Specifications

Specifications

Formula Weight 172.03
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Bromo-5-methylpyridine
Assay 97%
Molecular Formula C6H6BrN
MDL Number MFCD01646141
CAS 3430-16-8
Synonym 5-Bromo-3-picoline
Quantity 1g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.