Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromo-5-methylbenzeneboronic acid, 97%

$114.82 - $114.82

Chemical Identifiers

Chemical Name or Material 3-Bromo-5-methylbenzeneboronic acid
CAS 849062-36-8
Molecular Formula C7H8BBrO2
Formula Weight 214.85
MDL Number MFCD06411355
Synonym 3-Bromo-5-methylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5307003

  SDS
alfa aesar™
H5307003
1g Each for $114.82
Description & Specifications

Specifications

Formula Weight 214.85
Quantity 1g
Synonym 3-Bromo-5-methylphenylboronic acid
Health Hazard 1 Warning
CAS 849062-36-8
Assay 97%
Melting Point 276° to 278°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Bromo-5-methylbenzeneboronic acid
Molecular Formula C7H8BBrO2
MDL Number MFCD06411355
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.