Learn More
Alfa Aesar™ 3-Bromo-5-methylbenzeneboronic acid, 97%
Manufacturer: Alfa Aesar™ H5307006
Specifications
3-Bromo-5-methylbenzeneboronic acid | |
5g | |
C7H8BBrO2 | |
3-bromo-5-methylphenyl boronic acid, 3-bromo-5-methylbenzeneboronic acid, boronic acid,b-3-bromo-5-methylphenyl, 3-bromo-5-methylphenylboronicacid, acmc-209pyo, 3-bromo-5-methyl-phenyl boronic acid, 3-bromanyl-5-methyl-phenyl boronic acid | |
B(C1=CC(=CC(=C1)Br)C)(O)O | |
214.853 | |
214.85 |
276°C to 278°C | |
849062-36-8 | |
MFCD06411355 | |
KKEPYOBFWSGWLQ-UHFFFAOYSA-N | |
(3-bromo-5-methylphenyl)boronic acid | |
16217686 | |
97% |
Chemical Identifiers
849062-36-8 | |
214.853 | |
KKEPYOBFWSGWLQ-UHFFFAOYSA-N | |
16217686 | |
B(C1=CC(=CC(=C1)Br)C)(O)O |
C7H8BBrO2 | |
MFCD06411355 | |
3-bromo-5-methylphenyl boronic acid, 3-bromo-5-methylbenzeneboronic acid, boronic acid,b-3-bromo-5-methylphenyl, 3-bromo-5-methylphenylboronicacid, acmc-209pyo, 3-bromo-5-methyl-phenyl boronic acid, 3-bromanyl-5-methyl-phenyl boronic acid | |
(3-bromo-5-methylphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning